CA1 pyramidal neuron in a hippocampal slice filled with SBFI delivered from a patch pipette.
CA1 pyramidal neuron in a hippocampal slice filled with SBFI (Cat. no. S1262) delivered from a patch pipette (visible on the right). The image was obtained using two-photon excitation of SBFI at 790 nm. Image contributed by Christine R. Rose, Physiological Institute, University of Munich.
Related Products
Related Images
Cryostat section of mouse kidney. FluoCells® prepared slide #3, Alexa Fluor® 488 wheat germ agglutinin, Alexa Fluor® 568 phalloidin and DAPI. Go ›
Two-photon excitation imaging of calcium influx in a CA1 pyramidal cell spine Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›