Increase in accuracy and precision using the Qubit®
Accuracy and precision of the Qubit® Quantitation Platform. Ten replicates of lambda DNA at concentrations from 0.01 to 10 ng/µl were assayed using the Qubit® DNA HS Assay on the Qubit® 2.0 Fluorometer according to the standard kit protocol. The same concentrations of DNA were measured in 10 replicates using a NanoDrop® ND-1000 Spectrophotometer, and results were compared for both accuracy and precision. Each bar represents the average of 10 replicates. Error bars represent the standard deviations of the 10 replicates. The concentrations indicated are the concentrations of DNA in the starting samples, before dilution in the Qubit® assay tubes.