">
MSC-Advantage BSC
The Thermo Scientific MSC-Advantage Class II biological safety cabinet combines safety, energy efficiency and value for routine applications in the laboratory. The cabinet is energy efficient and provides both personal and product protection from biological hazards and contamination while affording users with a comfortable and ergonomic workspace. 
Value and performance combined
SmartClean window design
Front sash easily lowers to simplify cleaning, routine maintenance, and set up tasks.
SmartPort convenience
Easily pass tubing and wiring through sidewall to accommodate many application needs.
LED control panel
Large front-panel display (easily visible from seated position) provides constant readout of downflow and inflow velocities and filter usage status.
Features
1 SmartClean window – Easily access the interior of the biosafety cabinet by sliding down the front window. 
2 SmartPort convenience – Clean and safe passage of tubing and cables into the work area at sides of cabinet.
3 Customizable worktops – Choose from work surface designs that best suit your application and work style. Standard equipment includes two duplex receptacles and three service valves.
4 Dual DC Motors – Powering the SmartFlow airflow compensation system while delivering better energy efficiency and quieter operation.  Night setback mode furthers energy savings and system longevity.
5 Programmable UV light option – Extends bulb life, saves energy and reduces maintenance.
6 Ergonomic design – Sloped 10-degree front, arm rests and low noise enhance user comfort and productivity.
Applications
The MSC-Advantage biological safety cabinet is suitable for all applications where personnel, environmental and product protection is required, as well as protection from cross-contamination.
Typical markets: Academia and Clinical.
Typical applications are routine, research applications in cell culture, clinical research, life science research, and microbiology
Certifications
TÜV GS certified according to EN 12469 (all sizes)
NF Mark certified by LNE to EN 12469 and NF095 (0.9m and 1.2m models)
3D virtual tour
Equivalent product tour: 1300 Series BSC
360-degree, spin, zoom, rotate, discover and interact with our virtual 1300 Series BSC.
Product literature
Ordering information
Available in Europe and select Asian Pacific Countries.
Product
Size
See ordering details
MSC-Advantage BSC
0.9 m
51028225
1.2 m
51025411
1.5 m
51028226
1.8 m
51025413
">