Our techs are here to answer your questions, just email us.
Ask our techs ›
If you like this month's Q&A, check out our Talk Tech archive.
Question
Answer
The Thermo Scientific™ Series instruments include a built-in data logging capability.  The operator is allowed to create two different types of records, lrecs and srecs. Each record can contain up to 32 different fields or data items, and records can be created ranging from 1 to 60 minutes. This data can be easily accessed and downloaded through Thermo Scientific™ iPort software.
Refer to Data Logging Set Up Technical Bulletin for step-by-step instructions on how to set up data logging set up and how to download Thermo Scientific™ iPort software.
For addition additional information on Thermo Scientific™ iPort software, check out our November Fast5.
If you have additional questions, call the Technical Support team at (866) 282-0430 and select option 2.
tech-talk-showcase-image-230x195
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.
Subscribe to Talk Tech ›
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.