Primer Express Software allows you to design your own primers and probes using TaqMan and SYBR Green I dye chemistries for gene quantitation and allelic discrimination (SNP) real-time PCR applications. Developed specifically for use with our StepOne, StepOnePlus, 7300, 7500, 7500 Fast, 7900HT, ViiA 7, and QuantStudio real-time PCR systems, Primer Express Software provides customized application-specific documents for absolute⁄relative quantitation and allelic discrimination.
- Enables individual oligonucleotide design for real-time PCR applications.
- Supports assays based on TaqMan and SYBR Green I dye chemistries.
- Provides design flexibility, ease-of-use, and requires minimal optimization.
- Provides robust assay performance when used in accordance with Assay Development Guidelines
Software download
PC Download › Primer Express 3.0.1 Software Release Notes ›
For Research Use Only. Not for use in diagnostic procedures.