PeppermintStick phosphoprotein molecular weight standards separated on a 13% SDS polyacrylamide gel.
PeppermintStick phosphoprotein molecular weight standards (P33350, P27167) separated on a 13% SDS polyacrylamide gel. The markers contain (from largest to smallest) ß-galactosidase, bovine serum albumin (BSA), ovalbumin, ß-casein, avidin and lysozyme. Ovalbumin and ß-casein are phosphorylated. The gel was stained with Pro-Q Diamond phosphoprotein gel stain (blue) followed by SYPRO Ruby protein gel stain (red). The digital images were pseudocolored.