A zebrafish retina cryosection labeled with the mouse monoclonal antibody FRet 43 and detected using TSA Kit #2 with the HRP conjugate of goat anti–mouse IgG antibody and Alexa Fluor® 488 tyramide.
A zebrafish retina cryosection labeled with the mouse monoclonal antibody FRet 43 and detected using TSA Kit #2 with the HRP conjugate of goat anti–mouse IgG antibody and green-fluorescent Alexa Fluor® 488 tyramide (Cat. no. T20912). The nuclei were counterstained with blue-fluorescent Hoechst 33258 (Cat. no. H1398, H3569, H21491).