Bovine pulmonary artery endothelial cells (BPAEC) labeled with anti–OxPhos Complex IV subunit I antibody and detected using TSA Kit #4 with the HRP conjugate of goat anti–mouse IgG antibody and Alexa Fluor® 568 tyramide.
Fixed and permeabilized bovine pulmonary artery endothelial cells (BPAEC) labeled with anti–OxPhos Complex IV subunit I (human) antibody (anti–cytochrome oxidase subunit I) (Cat. no. A6403) and detected using TSA Kit #4 with the HRP conjugate of goat anti–mouse IgG antibody and Alexa Fluor® 568 tyramide (Cat. no. T20914).