ULYSIS reagent–labeled RNA hybridized to a microarray.
ULYSIS reagent–labeled RNA hybridized to a microarray. Poly(A)+ RNA samples from the spleen of an irradiated or unirradiated mouse were labeled using the Alexa Fluor® 594 ULS reagent (U21654) or Alexa Fluor® 546 ULS reagent (U21652), respectively. The labeled samples were mixed and hybridized to a cDNA microarray. The image was contributed by Rahul Mitra (Baylor College of Medicine) and Mini Kapoor, Thomas H. Burrows, and Rachel Grier (MD Anderson Cancer Center).
Related Products
Related Images
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›