Viable bovine pulmonary artery endothelial cells. JC-9.
A viable bovine pulmonary artery endothelial cell incubated with the ratiometric mitochondrial potential indicator, JC-9 (D22421). In live cells, JC-9 exists either as a green-fluorescent monomer at depolarized membrane potentials, or as a red-fluorescent J-aggregate at hyperpolarized membrane potentials.
Related Products
Related Images
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Agarose gel containing camptothecin-treated HL-60 cells. Go ›
Imaging autophagy in live HeLa cells with CellLight® reagents for mitochondria and lysosomes: Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›