Pyramidal cells of a mouse hippocampus and dentate gyrus. NeuroTrace® green fluorescent Nissl stain.
Pyramidal cells of the hippocampus and dentate gyrus in a transverse cryosection of paraformaldehyde-fixed mouse brain. NeuroTrace® green fluorescent Nissl stain (Cat. no. N21480) is localized to neuronal somata, while non-neuronal cells can be identified by the presence of DAPI-stained nuclei. This image is a composite of images taken using a 10× objective and filters appropriate for fluorescein and DAPI.
Related Products
Related Images
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Immunofluorescent Staining using Mouse HuC/HuD Neuronal Protein Monoclonal Antibody, Biotin-XX Conjugate. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›