Mouse fibroblasts. MitoTracker Red CMXRos, BODIPY FL phallacidin and DAPI.
Four-panel composite image of mouse fibroblasts that were incubated with MitoTracker Red CMXRos (M7512), and then formaldehyde-fixed, acetone-permeabilized and stained with the F-actin–specific probe, BODIPY FL phallacidin (B607) and with DAPI (D1306, D3571, D21490). Images were obtained by taking single and multiple exposures through bandpass optical filter sets appropriate for fluorescein, the Texas Red dye and DAPI.
Related Products
Related Images
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Endothelial cells. Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›