Peripheral nervous system of a Drosophila melanogaster embryo.
The peripheral nervous system of a wild-type (Canton-S) Drosophila melanogaster embryo labeled with the monoclonal 22c10 antibody (which detects a microtubule-associated protein) and subsequently visualized using green-fluorescent Alexa Fluor® 488 Rabbit Anti–Mouse IgG antibody (Cat. No. A11059). The actively dividing cells of the developing denticle bands were labeled with a rabbit anti–histone-H3 antibody and visualized using red-fluorescent Alexa Fluor® 594 Goat Anti–Rabbit IgG antibody (Cat. No. A11012). Finally, the nuclei, which are concentrated in the central nervous system, were counterstained with blue-fluorescent DAPI (Cat. No. D1306, D3571, D21490). Image contributed by Neville Cobbe, University of Edinburgh.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›