The Applied Biosystems VeritiPro Dx Thermal Cycler is an in vitro diagnostic (IVD) endpoint thermal cycler designed to amplify nucleic acids from human-derived specimens using polymerase chain reaction (PCR). It is manufactured to ISO 13485 and GMP requirements.
VeritiPro Dx Thermal Cycler conforms to:
- EU 2017/746 requirements and is CE- IVD labeled in Europe
- Classified as US-FDA class I medical device
- UK Medical devices regulations 2002 and is UKCA labeled in UK
This instrument delivers precise temperature control with a ramp rate of 6.0°C/sec. There is no need for protocol re-optimization when upgrading from Veriti Dx Thermal Cycler; the simulation mode supports seamless transition.
The VeritiPro Dx 96-well Thermal Cycler is only available for sale in the United States, Europe, and other selected countries. Contact your nearest sales representative for more details.
Key features of VeritiPro Dx: An in vitro diagnostic thermal cycler
The VeritiPro Dx Thermal Cycler features two security modes to provide user accessibility and safeguard the integrity of validated protocols and experimental results.
- Creation mode to enable users to see and open all folders to modify, view, and run all methods
- Run-only mode to restrict users to access the IVD folder, and only view and run IVD methods
Precise PCR optimization with VeriFlex block technology
VeriFlex Blocks on the VeritiPro Dx Thermal Cycler help provide precise control of your PCR optimization. With six independent Peltier blocks, you can precisely set and control the temperature in each block, helping give you maximal versatility, flexibility, and confidence in your diagnostic development.
- Run up to six precise and independent temperatures to determine the optimal annealing temperature for cycle sequencing or endpoint PCR
- VeriFlex Blocks maintain their thermal characteristics between optimizing and isothermal conditions, minimizing the need for further optimization steps
Learn more about precise PCR optimization with VeriFlex Blocks
Easy programming with an intuitive touch screen
The powerful yet simple-to-operate user interface on the VeritiPro Dx system is driven with a large, 8-inch color TFT LCD touch screen for editing or creating new methods and real-time run monitoring. A convenient USB port is also built into the VeritiPro Dx to simplify data transfer and software updates.
Thermo Fisher Scientific offers a comprehensive range of services and support to help keep your VeritiPro Dx Thermal Cycler's performance at peak level. The following services are available at no additional cost upon instrument purchase:
Basic package
- On-site installation qualification and operational qualification (IQ/OQ) services at the time of instrument installation
- Incidental post-repair IQ/OQ services
- Standard 1-year factory warranty
Service Contract Package
For labs operating in strictly regulated environments, the VeritiPro Dx Service Contract Package may be the ideal choice. The following service entitlements are additional to the basic package:
- Extended 1-year factory warranty
- Annual planned maintenance (PM) with OQ
The VeritiPro Dx Service Contract Package helps provide peace of mind with an extended warranty, annual testing, and documentation to help ensure that thermal cyclers in regulated environments have maximum uptime and produce high-quality data.
Qualification services
Instrument hardware qualifications for VeritiPro Dx Thermal Cycler include IQ/OQ services to verify and document the instrument's ability to meet manufacturer design specifications. Thermo Fisher Scientific's qualification specialists can partner with the customer to help deliver timely, cost-effective, and trusted qualification services that include reliable and audit-style documentation to help ensure thermal cyclers meet regulatory requirements.
Specifications
Block format | 96-well, 0.2 mL alloy block |
Maximum block ramp rate | 6.0°C/sec |
Temperature accuracy | ±0.25°C (35°C to 99.9°C) |
Temperature range | 0°C to 100°C |
Temperature uniformity | <0.5°C (30 sec after reaching 95°C) |
Reaction volume range | 10–100 µL |
Veriflex block range | 6 VeriFlex zones Supports: 25°C across block (5°C zone-to-zone) |
Display interface | 8 inch color TFT LCD touchscreen |
Instrument memory | USB, 8 GB on-board |
Number of programs | Up to 1,000 protocols |
Dimensions (L x W x H) | 46.5 x 24.5 x 21.7 cm (18.3 in x 9.8 in x 8.5 in) |
Weight | 12 kg (26.5 lbs) |
Voltage | 100–120 V; 200–240 V; 50–60 Hz |
Noise output | Operation <53 dB(A) Idling <40 dB(A) |
Sustainability | Sleep (Standby mode) to reduce power consumption compared to idling mode |
Plastics compatibility chart for Applied Biosystems instruments
PCR and cDNA Synthesis Support Center
Tips and troubleshooting help
School of Molecular Biology, PCR Education
Technical content for new and experienced molecular biologists
Thermal Cycler Software and Firmware Files
Download the latest updates for your instrument
Interested in private label thermal cyclers?
Submit an inquiry
Service
We provide a full range of service options for all Applied Biosystems thermal cyclers.
View service plans
Instrument hardware qualification service
Have one of our engineers perform temperature qualifications at your site ›
*The VeritiPro Dx Thermal Cycler is available for in vitro diagnostic use in the United States of America and Europe and other selected countries only. The customer is responsible for any validation of assays, reagents, and consumables, and compliance with any regulatory requirements that pertain to their procedures and instrument use.