Transportable area gas monitors are ideal for routine plant maintenance, shutdown work, commissioning, tank farms, drilling rigs, disaster response and fence-line monitoring, enabling people to work with confidence in hazardous areas. Simply place the monitor nearby and it will alert workers to area gas threats so they can take action.
*Please note these products are available in Australia only.
Honeywell BW™ RigRat |
Blackline G7 EXO |
---|---|
Up to 6 gas sensors, pumped or diffusion | Up to 5 gas sensors, pumped or diffusion |
Choose from sensors for toxic gases, oxygen, and combustible (LEL) gases; PID detector for VOCs; IR detector for flammable gases | Choose from sensors for toxic gases, oxygen, and combustible (LEL) gases; PID detector for VOCs; IR detector for flammable gases |
Up to 8 weeks of run time | Up to 90 days of run time |
Battery, mains power, or solar power options, with intrinsically safe line power supply input | Battery, mains power, or solar power options, with intrinsically safe line power supply input |
Simple setup via Bluetooth to Smartphone | Cloud connected setup, via 3G/4G Network |
Wireless mesh-connection to other RigRat for complete area monitoring | Wireless cloud-connection to other EXO for complete area monitoring |
ATEX/IECEx Zone 0 rating | ATEX/IECEx Zone 0 rating |
Optional sensors:
|
Optional satellite connection |
One button operation | Three-button keypad |
Connect to wireless portable gas monitors via BLE. Connect to site controller, along with other fixed gas instruments |
SOS alert latch and two-way voice calling to live monitoring team, via 3G/4G network |
Analog and relay outputs for external alarm control | Analog and relay outputs for external alarm control |