Fibrous materials, such as carbon or glass fiber (CFRP, GFRP), fabric (multi-layer composites), fiber-reinforced concrete (FRC), and organic fibers are used in a wide range of applications and industries (automotive, aerospace, etc.). Imaging techniques such as microCT or SEM allow for analysis of those materials; for example, to detect manufacturing imperfections or to quantify microstructures to obtain insights on mechanical properties (strength, stiffness, etc.).
Thermo Scientific Avizo Software is an all-in-one image analysis platform that allows the visualization, processing, and quantification of fibrous materials. Avizo Software enables the detection of manufacturing process damage such as matrix cracking, fiber/matrix debonding, or fiber breakage. Imperfections in the material can be quantified locally (voids, cracks) or globally (cross-sectional distortions or misalignments).
Avizo Software provides dedicated tools for fiber analysis:
- Automatic segmentation of individual high-resolution fibers
- Major orientations of low-resolution fibers
- Quantification such as contact point or distance to closest neighbor
- Fiber shape statistics (diameter, length, tortuosity, area, volume fraction, cross section perimeter, and area)
- Statistics relative to the fibers' density (volume fraction, area, and tensor per material subdomain)
- 3D plot of fiber's orientation on a sphere or half-sphere with properties mapped as color and height
- Advanced fiber filtering modules