LabLink QC software streamlines your daily QC workflow by enabling instant updates of your QC testing data and lets you compare your data with other or in affiliated labs over the cloud.
Features and benefits
Troubleshoot faster with real-time peer comparison
- Compare your data with labs around the globe or just your affiliated network.
- Determine if a worrisome result is an instrument problem, widespread issue, or acceptable value.
- Peer data is updated in real-time, so you can identify trends and shifts sooner.
Eliminate errors with automatic data entry
- Automatically send QC data from your instrument, L.I.S., or middleware to LabLink xL.
- Say goodbye to manual entry and submission cutoff dates.
Stay "in control" with audit-friendly reports
- Easy-to-read reports help you monitor individual lab performance by shift, day, and month.
- View peer data alongside your lab's data.
- Reports are automatically delivered to your email inbox.
- Review and digitally sign with LabLink xL Signature Verification.
Sigma score & method performance
The new method performance report gives a sigma score for each instrument.
- See which instruments are performing the best for a particular method and test.
- Choose between CLIA quality goals or custom goals for total allowable error.
Day-to-day QC is all in one place
- Perform all your daily QC activities inside LabLink xL.
- Compare Levey-Jennings Charts for multiple instruments.
- Select Westgard and Rilibak violation rules.
- Flag and comment on individual data points.
- Receive updated package inserts and material safety data sheets.
Easy to setup
Use with MAS products or any other QC vendor**
Truly real-time
calculations
Audit Friendly
Reports
*LabLink xL only requires an internet browser to view the interface. Some autoconnectivity options may require software installation.
**Non-MAS products are supported by our autoconnectivity and Daily QC features. Peer data for non-MAS products is not available at this time.
**Non-MAS products are supported by our autoconnectivity and Daily QC features. Peer data for non-MAS products is not available at this time.