Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: SYT17.
Plays a role in dendrite formation by melanocytes. Classified as a brain and kidney protein (not to be confused with the BigK K+ channel); may play a role in cAMP mediated signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B/K protein; brain and kidney protein; Protein B/K; Synaptotagmin XVII; Synaptotagmin-17; SytXVII
Gene Aliases: Bk; SYT17; sytXVII
UniProt ID: (Human) Q9BSW7, (Rat) Q62807, (Mouse) Q920M7
Entrez Gene ID: (Human) 51760, (Rat) 192189, (Mouse) 110058
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support