Product References
Identification of Tau protein as a novel marker for maturation and pathological changes of oligodendrocytes.
Glia
Torii T,Miyamoto Y,Nakata R,Higashi Y,Shinmyo Y,Kawasaki H,Miyasaka T,Misonou H
Sat Apr 01 00:00:00 EDT 2023
Remyelination in neuromyelitis optica spectrum disorder is promoted by edaravone through mTORC1 signaling activation.
Glia
Luo W,Xu H,Xu L,Jiang W,Chen C,Chang Y,Liu C,Tian Z,Qiu X,Xie C,Li X,Chen H,Lai S,Wu L,Cui Y,Tang C,Qiu W
Wed Feb 01 00:00:00 EST 2023
XLF/Cernunnos loss impairs mouse brain development by altering symmetric proliferative divisions of neural progenitors.
Cell reports
Bery A,Etienne O,Mouton L,Mokrani S,Granotier-Beckers C,Gauthier LR,Feat-Vetel J,Kortulewski T,Pérès EA,Desmaze C,Lestaveal P,Barroca V,Laugeray A,Boumezbeur F,Abramovski V,Mortaud S,Menuet A,Le Bihan D,Villartay JP,Boussin FD
Tue Apr 25 00:00:00 EDT 2023
Molecules of senescent glial cells differentiate Alzheimer's disease from ageing.
Journal of neurology, neurosurgery, and psychiatry
Dai L,Gao F,Wang Q,Lv X,Cheng Z,Wu Y,Chai X,Zetterberg H,Blennow K,Levey AI,Shi J,Shen Y
Sat Jul 01 00:00:00 EDT 2023
Baicalin facilitates remyelination and suppresses neuroinflammation in rats with chronic cerebral hypoperfusion by activating Wnt/β-catenin and inhibiting NF-κB signaling.
Behavioural brain research
Xiao Y,Guan T,Yang X,Xu J,Zhang J,Qi Q,Teng Z,Dong Y,Gao Y,Li M,Meng N,Lv P
Tue Mar 28 00:00:00 EDT 2023
Baicalin facilitates remyelination and suppresses neuroinflammation in rats with chronic cerebral hypoperfusion by activating Wnt/β-catenin and inhibiting NF-κB signaling.
Behavioural brain research
Xiao Y,Guan T,Yang X,Xu J,Zhang J,Qi Q,Teng Z,Dong Y,Gao Y,Li M,Meng N,Lv P
Tue Mar 28 00:00:00 EDT 2023
Choroid Plexus Modulates Subventricular Zone Adult Neurogenesis and Olfaction Through Secretion of Small Extracellular Vesicles.
bioRxiv : the preprint server for biology
Liu LL,Shannahan J,Zheng W
Sat Mar 18 00:00:00 EDT 2023
Suppression of premature transcription termination leads to reduced mRNA isoform diversity and neurodegeneration.
Neuron
LaForce GR,Farr JS,Liu J,Akesson C,Gumus E,Pinkard O,Miranda HC,Johnson K,Sweet TJ,Ji P,Lin A,Coller J,Philippidou P,Wagner EJ,Schaffer AE
Wed Apr 20 00:00:00 EDT 2022
Variants of the adeno-associated virus serotype 9 with enhanced penetration of the blood-brain barrier in rodents and primates.
Nature biomedical engineering
Yao Y,Wang J,Liu Y,Qu Y,Wang K,Zhang Y,Chang Y,Yang Z,Wan J,Liu J,Nakashima H,Lawler SE,Chiocca EA,Cho CF,Bei F
Tue Nov 01 00:00:00 EDT 2022
Gut microbiota shapes social dominance through modulating HDAC2 in the medial prefrontal cortex.
Cell reports
Wang T,Xu J,Xu Y,Xiao J,Bi N,Gu X,Wang HL
Tue Mar 08 00:00:00 EST 2022
Bovine intracranial neoplasia: A retrospective case series.
Veterinary pathology
Jahns H,McElroy MC
Thu Sep 01 00:00:00 EDT 2022
RSL3 triggers glioma stem cell differentiation via the Tgm2/AKT/ID1 signaling axis.
Biochimica et biophysica acta. Molecular basis of disease
Li M,Song D,Chen X,Wang X,Xu L,Yang M,Yang J,Kalvakolanu DV,Wei X,Liu X,Li Y,Guo B,Zhang L
Thu Dec 01 00:00:00 EST 2022
RSL3 triggers glioma stem cell differentiation via the Tgm2/AKT/ID1 signaling axis.
Biochimica et biophysica acta. Molecular basis of disease
Li M,Song D,Chen X,Wang X,Xu L,Yang M,Yang J,Kalvakolanu DV,Wei X,Liu X,Li Y,Guo B,Zhang L
Thu Dec 01 00:00:00 EST 2022
RSL3 triggers glioma stem cell differentiation via the Tgm2/AKT/ID1 signaling axis.
Biochimica et biophysica acta. Molecular basis of disease
Li M,Song D,Chen X,Wang X,Xu L,Yang M,Yang J,Kalvakolanu DV,Wei X,Liu X,Li Y,Guo B,Zhang L
Thu Dec 01 00:00:00 EST 2022
RSL3 triggers glioma stem cell differentiation via the Tgm2/AKT/ID1 signaling axis.
Biochimica et biophysica acta. Molecular basis of disease
Li M,Song D,Chen X,Wang X,Xu L,Yang M,Yang J,Kalvakolanu DV,Wei X,Liu X,Li Y,Guo B,Zhang L
Thu Dec 01 00:00:00 EST 2022
Neuron-specific ablation of the Krabbe disease gene galactosylceramidase in mice results in neurodegeneration.
PLoS biology
Kreher C,Favret J,Weinstock NI,Maulik M,Hong X,Gelb MH,Wrabetz L,Feltri ML,Shin D
Fri Jul 01 00:00:00 EDT 2022
Stemazole Promotes Oligodendrocyte Precursor Cell Survival In Vitro and Remyelination In Vivo.
International journal of molecular sciences
Zhu Y,Chen M,Zhang Y,Han M
Thu Sep 15 00:00:00 EDT 2022
The Epigenetic Regulation of OLIG2 by Histone Demethylase KDM6B in Glioma Cells.
Journal of molecular neuroscience : MN
Sui A,Han B,Ren W,He W,Gao C,Han X,Liu S,Zhang Y,Qi X,Guo X
Sun May 01 00:00:00 EDT 2022
Clemastine Promotes Differentiation of Oligodendrocyte Progenitor Cells Through the Activation of ERK1/2 via Muscarinic Receptors After Spinal Cord Injury.
Frontiers in pharmacology
Tong LY,Deng YB,Du WH,Zhou WZ,Liao XY,Jiang X
Sat Jul 23 00:00:00 EDT 2022
Tetrahydrofolate Alleviates the Inhibitory Effect of Oxidative Stress on Neural Stem Cell Proliferation through PTEN/Akt/mTOR Pathway.
Oxidative medicine and cellular longevity
Zhang X,Liu Z,Yang W,Zhao F,Zhang C,Feng H,Zhou T,Zhong J,Zou Y,Feng H,Ge H,Hu R
Thu Mar 24 00:00:00 EDT 2022
Oligodendrocyte differentiation alters tRNA modifications and codon optimality-mediated mRNA decay.
Nature communications
Martin S,Allan KC,Pinkard O,Sweet T,Tesar PJ,Coller J
Thu Aug 25 00:00:00 EDT 2022
TGFBI secreted by tumor-associated macrophages promotes glioblastoma stem cell-driven tumor growth via integrin αvβ5-Src-Stat3 signaling.
Theranostics
Peng P,Zhu H,Liu D,Chen Z,Zhang X,Guo Z,Dong M,Wan L,Zhang P,Liu G,Zhang S,Dong F,Hu F,Cheng F,Huang S,Guo D,Zhang B,Yu X,Wan F
Thu Jun 09 00:00:00 EDT 2022
Alpha-Asaronol Alleviates Dysmyelination by Enhancing Glutamate Transport Through the Activation of PPARγ-GLT-1 Signaling in Hypoxia-Ischemia Neonatal Rats.
Frontiers in pharmacology
Ge Y,Zhen F,Liu Z,Feng Z,Wang G,Zhang C,Wang X,Sun Y,Zheng X,Bai Y,Yao R
Wed Apr 13 00:00:00 EDT 2022
Matrine inhibits the Wnt3a/β-catenin/TCF7L2 signaling pathway in experimental autoimmune encephalomyelitis.
Journal of neuroimmunology
Ma R,Chu Y,Dou M,Jing Y,Zhao X,Li X,Li S,Zhang Y,Wang L,Zhu L
Wed Jun 15 00:00:00 EDT 2022
PTBP-1 and TNF-α/NF-κB are involved in repair mechanisms of human umbilical cord mesenchymal stem cell transplantation in mice with spinal cord injury.
American journal of translational research
Cao H,Ji X,Wang Q,Guan X,Wei W,Li Y,Zou W,Liu J
Sat Aug 13 00:00:00 EDT 2022
PTBP-1 and TNF-α/NF-κB are involved in repair mechanisms of human umbilical cord mesenchymal stem cell transplantation in mice with spinal cord injury.
American journal of translational research
Cao H,Ji X,Wang Q,Guan X,Wei W,Li Y,Zou W,Liu J
Sat Aug 13 00:00:00 EDT 2022
Non-canonical Targets of HIF1a Impair Oligodendrocyte Progenitor Cell Function.
Cell stem cell
Allan KC,Hu LR,Scavuzzo MA,Morton AR,Gevorgyan AS,Cohn EF,Clayton BLL,Bederman IR,Hung S,Bartels CF,Madhavan M,Tesar PJ
Thu Feb 04 00:00:00 EST 2021
Molecular Features of the Measles Virus Viral Fusion Complex That Favor Infection and Spread in the Brain.
mBio
Mathieu C,Bovier FT,Ferren M,Lieberman NAP,Predella C,Lalande A,Peddu V,Lin MJ,Addetia A,Patel A,Outlaw V,Corneo B,Dorrello NV,Briese T,Hardie D,Horvat B,Moscona A,Greninger AL,Porotto M
Tue Jun 29 00:00:00 EDT 2021
A five-m6A regulatory gene signature is a prognostic biomarker in lung adenocarcinoma patients.
Aging
Wu X,Sheng H,Wang L,Xia P,Wang Y,Yu L,Lv W,Hu J
Fri Mar 26 00:00:00 EDT 2021
Transgenic Mice Expressing Human α-Synuclein 1-103 Fragment as a Novel Model of Parkinson's Disease.
Frontiers in aging neuroscience
Tian Y,He M,Pan L,Yuan X,Xiong M,Meng L,Yao Z,Yu Z,Ye K,Zhang Z
Wed Nov 10 00:00:00 EST 2021
Melatonin pretreatment alleviates the long-term synaptic toxicity and dysmyelination induced by neonatal Sevoflurane exposure via MT1 receptor-mediated Wnt signaling modulation.
Journal of pineal research
Liang L,Zeng T,Zhao Y,Lu R,Guo B,Xie R,Tang W,Zhang L,Mao Z,Yang X,Wu S,Wang Y,Zhang H
Wed Dec 01 00:00:00 EST 2021
ADAM10 suppresses demyelination and reduces seizure susceptibility in cuprizone-induced demyelination model.
Free radical biology & medicine
Zhu X,Yao Y,Yang J,Zhang C,Li X,Zhang A,Liu X,Zhang C,Gan G
Sun Aug 01 00:00:00 EDT 2021
Maternal high sugar and fat diet benefits offspring brain function via targeting on the gut-brain axis.
Aging
Wang D,Zhang H,Zeng M,Tang X,Zhu X,Guo Y,Qi L,Xie Y,Zhang M,Chen D
Fri Mar 26 00:00:00 EDT 2021
Phospholipase D and phosphatidylinositol-4-phosphate 5-kinase 1 are involved in the regulation of oligodendrocyte morphological differentiation.
Experimental cell research
Kato Y,Ochiai A,Seki Y,Morimoto T,Oizumi H,Ohbuchi K,Mizoguchi K,Yamamoto M,Sakagami H,Miyamoto Y,Yamauchi J
Sun Aug 01 00:00:00 EDT 2021
Giant ankyrin-B mediates transduction of axon guidance and collateral branch pruning factor sema 3A.
eLife
Creighton BA,Afriyie S,Ajit D,Casingal CR,Voos KM,Reger J,Burch AM,Dyne E,Bay J,Huang JK,Anton ES,Fu MM,Lorenzo DN
Tue Nov 23 00:00:00 EST 2021
TREM2 overexpression rescues cognitive deficits in APP/PS1 transgenic mice by reducing neuroinflammation via the JAK/STAT/SOCS signaling pathway.
Experimental neurology
Ruganzu JB,Zheng Q,Wu X,He Y,Peng X,Jin H,Zhou J,Ma R,Ji S,Ma Y,Qian Y,Wang Y,Yang W
Mon Feb 01 00:00:00 EST 2021
Multiple sclerosis risk gene Mertk is required for microglial activation and subsequent remyelination.
Cell reports
Shen K,Reichelt M,Kyauk RV,Ngu H,Shen YA,Foreman O,Modrusan Z,Friedman BA,Sheng M,Yuen TJ
Tue Mar 09 00:00:00 EST 2021
Multiple sclerosis risk gene Mertk is required for microglial activation and subsequent remyelination.
Cell reports
Shen K,Reichelt M,Kyauk RV,Ngu H,Shen YA,Foreman O,Modrusan Z,Friedman BA,Sheng M,Yuen TJ
Tue Mar 09 00:00:00 EST 2021
Macrophages Expressing GALC Improve Peripheral Krabbe Disease by a Mechanism Independent of Cross-Correction.
Neuron
Weinstock NI,Shin D,Dhimal N,Hong X,Irons EE,Silvestri NJ,Reed CB,Nguyen D,Sampson O,Cheng YC,Lau JTY,Bongarzone ER,Kofler J,Escolar ML,Gelb MH,Wrabetz L,Feltri ML
Wed Jul 08 00:00:00 EDT 2020
Dl-3-n-Butylphthalide Promotes Remyelination and Suppresses Inflammation by Regulating AMPK/SIRT1 and STAT3/NF-κB Signaling in Chronic Cerebral Hypoperfusion.
Frontiers in aging neuroscience
Li M,Meng N,Guo X,Niu X,Zhao Z,Wang W,Xie X,Lv P
Mon Sep 28 00:00:00 EDT 2020
Dl-3-n-Butylphthalide Promotes Remyelination and Suppresses Inflammation by Regulating AMPK/SIRT1 and STAT3/NF-κB Signaling in Chronic Cerebral Hypoperfusion.
Frontiers in aging neuroscience
Li M,Meng N,Guo X,Niu X,Zhao Z,Wang W,Xie X,Lv P
Mon Sep 28 00:00:00 EDT 2020
Deficiency of microglial Hv1 channel is associated with activation of autophagic pathway and ROS production in LPC-induced demyelination mouse model.
Journal of neuroinflammation
Chen M,Yang LL,Hu ZW,Qin C,Zhou LQ,Duan YL,Bosco DB,Wu LJ,Zhan KB,Xu SB,Tian DS
Fri Nov 06 00:00:00 EST 2020
Suppression of proteolipid protein rescues Pelizaeus-Merzbacher disease.
Nature
Elitt MS,Barbar L,Shick HE,Powers BE,Maeno-Hikichi Y,Madhavan M,Allan KC,Nawash BS,Gevorgyan AS,Hung S,Nevin ZS,Olsen HE,Hitomi M,Schlatzer DM,Zhao HT,Swayze A,LePage DF,Jiang W,Conlon RA,Rigo F,Tesar PJ
Tue Sep 01 00:00:00 EDT 2020
Leptin Receptor Deficiency Protects Mice against Chronic Cerebral Hypoperfusion-Induced Neuroinflammation and White Matter Lesions.
Mediators of inflammation
Du Y,Song Y,Zhang X,Luo Y,Zou W,Zhang J,Fu J
Tue Sep 21 00:00:00 EDT 2021
Requirements of LEFTY and Nodal overexpression for tumor cell survival under hypoxia in glioblastoma.
Molecular carcinogenesis
Matsumoto T,Chino H,Akiya M,Hashimura M,Yokoi A,Tochimoto M,Nakagawa M,Jiang Z,Saegusa M
Tue Dec 01 00:00:00 EST 2020
Requirements of LEFTY and Nodal overexpression for tumor cell survival under hypoxia in glioblastoma.
Molecular carcinogenesis
Matsumoto T,Chino H,Akiya M,Hashimura M,Yokoi A,Tochimoto M,Nakagawa M,Jiang Z,Saegusa M
Tue Dec 01 00:00:00 EST 2020
Distribution of endogenous normal tau in the mouse brain.
The Journal of comparative neurology
Kubo A,Misonou H,Matsuyama M,Nomori A,Wada-Kakuda S,Takashima A,Kawata M,Murayama S,Ihara Y,Miyasaka T
Mon Apr 01 00:00:00 EDT 2019
Distribution of endogenous normal tau in the mouse brain.
The Journal of comparative neurology
Kubo A,Misonou H,Matsuyama M,Nomori A,Wada-Kakuda S,Takashima A,Kawata M,Murayama S,Ihara Y,Miyasaka T
Mon Apr 01 00:00:00 EDT 2019
Myelin Damage in Diffuse Axonal Injury.
Frontiers in neuroscience
Mu J,Li M,Wang T,Li X,Bai M,Zhang G,Kong J
Wed Sep 30 00:00:00 EDT 2020
Valproic Acid Enhances Reprogramming Efficiency and Neuronal Differentiation on Small Molecules Staged-Induction Neural Stem Cells: Suggested Role of mTOR Signaling.
Frontiers in neuroscience
Duan Q,Li S,Wen X,Sunnassee G,Chen J,Tan S,Guo Y
Wed Sep 30 00:00:00 EDT 2020
Restorative effect of modified dioscorea pills on the structure of hippocampal neurovascular unit in an animal model of chronic cerebral hypoperfusion.
Heliyon
Li HB
Mon Apr 01 00:00:00 EDT 2019
Chondroitin sulfate proteoglycan represses neural stem/progenitor cells migration via PTPσ/α-actinin4 signaling pathway.
Journal of cellular biochemistry
Zhong J,Lan C,Zhang C,Yang Y,Chen WX,Zhang KY,Zhao HL,Fang XY,Li HH,Tan L,Wang P,Ge HF,Hu R,Feng H
Mon Jul 01 00:00:00 EDT 2019
G protein-coupled estrogen receptor 1 negatively regulates the proliferation of mouse-derived neural stem/progenitor cells via extracellular signal-regulated kinase pathway.
Brain research
Zhong J,Ge HF,Zhang C,Chen JY,Li HH,Fang XY,Tan L,Liu X,Jia ZC,Feng H,Hu R
Mon Jul 01 00:00:00 EDT 2019
Transplantation of hypoxic preconditioned neural stem cells benefits functional recovery via enhancing neurotrophic secretion after spinal cord injury in rats.
Journal of cellular biochemistry
Fan WL,Liu P,Wang G,Pu JG,Xue X,Zhao JH
Fri Jun 01 00:00:00 EDT 2018
Transplantation of hypoxic preconditioned neural stem cells benefits functional recovery via enhancing neurotrophic secretion after spinal cord injury in rats.
Journal of cellular biochemistry
Fan WL,Liu P,Wang G,Pu JG,Xue X,Zhao JH
Fri Jun 01 00:00:00 EDT 2018
Examining the role of the surfactant family member SFTA3 in interneuron specification.
PloS one
Chen CY,Anderson NC,Becker S,Schicht M,Stoddard C,Bräuer L,Paulsen F,Grabel L
Wed Apr 10 00:00:00 EDT 2019
Examining the role of the surfactant family member SFTA3 in interneuron specification.
PloS one
Chen CY,Anderson NC,Becker S,Schicht M,Stoddard C,Bräuer L,Paulsen F,Grabel L
Wed Apr 10 00:00:00 EDT 2019
Rapid functional genetics of the oligodendrocyte lineage using pluripotent stem cells.
Nature communications
Lager AM,Corradin OG,Cregg JM,Elitt MS,Shick HE,Clayton BLL,Allan KC,Olsen HE,Madhavan M,Tesar PJ
Thu Sep 13 00:00:00 EDT 2018
Rapid functional genetics of the oligodendrocyte lineage using pluripotent stem cells.
Nature communications
Lager AM,Corradin OG,Cregg JM,Elitt MS,Shick HE,Clayton BLL,Allan KC,Olsen HE,Madhavan M,Tesar PJ
Thu Sep 13 00:00:00 EDT 2018
Antibody incubation at 37°C improves fluorescent immunolabeling in free-floating thick tissue sections.
BioTechniques
Xiao X,Feng YP,Du B,Sun HR,Ding YQ,Qi JG
Wed Mar 01 00:00:00 EST 2017
Antibody incubation at 37°C improves fluorescent immunolabeling in free-floating thick tissue sections.
BioTechniques
Xiao X,Feng YP,Du B,Sun HR,Ding YQ,Qi JG
Wed Mar 01 00:00:00 EST 2017
Resetting cancer stem cell regulatory nodes upon MYC inhibition.
EMBO reports
Galardi S,Savino M,Scagnoli F,Pellegatta S,Pisati F,Zambelli F,Illi B,Annibali D,Beji S,Orecchini E,Alberelli MA,Apicella C,Fontanella RA,Michienzi A,Finocchiaro G,Farace MG,Pavesi G,Ciafrè SA,Nasi S
Thu Dec 01 00:00:00 EST 2016
Resetting cancer stem cell regulatory nodes upon MYC inhibition.
EMBO reports
Galardi S,Savino M,Scagnoli F,Pellegatta S,Pisati F,Zambelli F,Illi B,Annibali D,Beji S,Orecchini E,Alberelli MA,Apicella C,Fontanella RA,Michienzi A,Finocchiaro G,Farace MG,Pavesi G,Ciafrè SA,Nasi S
Thu Dec 01 00:00:00 EST 2016
Formylpeptide Receptors Promote the Migration and Differentiation of Rat Neural Stem Cells.
Scientific reports
Wang G,Zhang L,Chen X,Xue X,Guo Q,Liu M,Zhao J
Fri May 13 00:00:00 EDT 2016
Formylpeptide Receptors Promote the Migration and Differentiation of Rat Neural Stem Cells.
Scientific reports
Wang G,Zhang L,Chen X,Xue X,Guo Q,Liu M,Zhao J
Fri May 13 00:00:00 EDT 2016
Poly-L-ornithine enhances migration of neural stem/progenitor cells via promoting α-Actinin 4 binding to actin filaments.
Scientific reports
Ge H,Yu A,Chen J,Yuan J,Yin Y,Duanmu W,Tan L,Yang Y,Lan C,Chen W,Feng H,Hu R
Tue Nov 22 00:00:00 EST 2016
Poly-L-ornithine enhances migration of neural stem/progenitor cells via promoting α-Actinin 4 binding to actin filaments.
Scientific reports
Ge H,Yu A,Chen J,Yuan J,Yin Y,Duanmu W,Tan L,Yang Y,Lan C,Chen W,Feng H,Hu R
Tue Nov 22 00:00:00 EST 2016
Poly-L-ornithine promotes preferred differentiation of neural stem/progenitor cells via ERK signalling pathway.
Scientific reports
Ge H,Tan L,Wu P,Yin Y,Liu X,Meng H,Cui G,Wu N,Lin J,Hu R,Feng H
Tue Oct 27 00:00:00 EDT 2015
Poly-L-ornithine promotes preferred differentiation of neural stem/progenitor cells via ERK signalling pathway.
Scientific reports
Ge H,Tan L,Wu P,Yin Y,Liu X,Meng H,Cui G,Wu N,Lin J,Hu R,Feng H
Tue Oct 27 00:00:00 EDT 2015
Poly-L-ornithine promotes preferred differentiation of neural stem/progenitor cells via ERK signalling pathway.
Scientific reports
Ge H,Tan L,Wu P,Yin Y,Liu X,Meng H,Cui G,Wu N,Lin J,Hu R,Feng H
Tue Oct 27 00:00:00 EDT 2015
Poly-L-ornithine promotes preferred differentiation of neural stem/progenitor cells via ERK signalling pathway.
Scientific reports
Ge H,Tan L,Wu P,Yin Y,Liu X,Meng H,Cui G,Wu N,Lin J,Hu R,Feng H
Tue Oct 27 00:00:00 EDT 2015
Neuropeptide Y has a protective role during murine retrovirus-induced neurological disease.
Journal of virology
Du M,Butchi NB,Woods T,Morgan TW,Peterson KE
Mon Nov 01 00:00:00 EDT 2010
Neuropeptide Y has a protective role during murine retrovirus-induced neurological disease.
Journal of virology
Du M,Butchi NB,Woods T,Morgan TW,Peterson KE
Mon Nov 01 00:00:00 EDT 2010