Product References
Inhibition of TREM-1 ameliorates Lipopolysaccharide-induced depressive-like behaviors by alleviating neuroinflammation in the PFC via PI3K/Akt signaling pathway.
Behavioural brain research
Fu A,Qiao F,Feng H,Luo Q
Sun Jul 09 00:00:00 EDT 2023
Photobiomodulation treatment inhibits neurotoxic astrocytic polarization and protects neurons in in vitro and in vivo stroke models.
Neurochemistry international
Feng Y,Yang L,Ma X,Huang Z,Zong X,Citadin CT,Lin HW,Zhang Q
Sun Jan 01 00:00:00 EST 2023
Methylene blue targets PHD3 expression in murine microglia to mitigate lipopolysaccharide-induced neuroinflammation and neurocognitive impairments.
International immunopharmacology
Ou G,Che J,Dong J,Deng Y,Jiang X,Sun Y,He Z,Chen W,Zhang J
Sat Jul 01 00:00:00 EDT 2023
Daphnetin alleviates neuropathic pain in chronic constrictive injury rats via regulating the NF-κB dependent CXCL1/CXCR2 signaling pathway.
Pharmaceutical biology
Zhang T,Liang W,Ou W,Zhang M,Cui S,Zhang S
Fri Dec 01 00:00:00 EST 2023
Hydrogen Sulfide Inhibited Sympathetic Activation in D-Galactose-Induced Aging Rats by Upregulating Klotho and Inhibiting Inflammation in the Paraventricular Nucleus.
Biomedicines
Yu H,Yu Q,Mi Y,Wang P,Jin S,Xiao L,Guo Q,Wu Y
Wed Feb 15 00:00:00 EST 2023
Hydroxysafflor yellow a confers neuroprotection against acute traumatic brain injury by modulating neuronal autophagy to inhibit NLRP3 inflammasomes.
Journal of ethnopharmacology
Lai Z,Li C,Ma H,Hua S,Liu Z,Huang S,Liu K,Li J,Feng Z,Cai Y,Zou Y,Tang Y,Jiang X
Tue May 23 00:00:00 EDT 2023
Inhibiting PAC1 receptor internalization and endosomal ERK pathway activation may ameliorate hyperalgesia in a chronic migraine rat model.
Cephalalgia : an international journal of headache
Zhang L,Zhou Y,Wang Y,Yang L,Wang Y,Shan Z,Liang J,Xiao Z
Sat Apr 01 00:00:00 EDT 2023
Epimedium Aqueous Extract Ameliorates Cerebral Ischemia/Reperfusion Injury through Inhibiting ROS/NLRP3-Mediated Pyroptosis.
Antioxidants (Basel, Switzerland)
Wu X,Wei J,Yi Y,Shu G,He Z,Gong Q,Gao J
Tue Apr 25 00:00:00 EDT 2023
Negative regulation of TREM2-mediated C9orf72 poly-GA clearance by the NLRP3 inflammasome.
Cell reports
Shu X,Wei C,Tu WY,Zhong K,Qi S,Wang A,Bai L,Zhang SX,Luo B,Xu ZZ,Zhang K,Shen C
Tue Feb 28 00:00:00 EST 2023
Tonic Meningeal Interleukin-10 Upregulates Delta Opioid Receptor to Prevent Relapse to Pain.
bioRxiv : the preprint server for biology
Inyang KE,Sim J,Clark KB,Matan G,Monahan K,Evans C,Beng P,Ma JV,Heijnen CJ,Dantzer R,Scherrer G,Kavelaars A,Bernard M,Aldhamen Y,Folger JK,Laumet G
Fri Jun 09 00:00:00 EDT 2023
Activation of the microglial P2X7R/NLRP3 inflammasome mediates central sensitization in a mouse model of medication overuse headache.
Frontiers in molecular neuroscience
Wang Y,Dong L,Zhang Y,Zhang Y,Qin G,Zhang D,Chen L,He W,Zhou J
Sat Jul 01 00:00:00 EDT 2023
Peroxiredoxin 2 Is a Potential Objective Indicator for Severity and the Clinical Status of Subarachnoid Hemorrhage Patients.
Disease markers
Peng Z,Pang C,Li XJ,Zhang HS,Zhang JT,Zhu Q,Xu HJ,Gao YY,Zhuang Z,Li W,Zhang QR,Lu Y,Hang CH
Fri Feb 17 00:00:00 EST 2023
Study on Neuroprotective Mechanism of Houshiheisan in Ischemic Stroke Based on Transcriptomics and Experimental Verification.
Evidence-based complementary and alternative medicine : eCAM
Cheng H,Zhang Y,Guo X,Wang X,Wang H,Zhao H,Wang L,Zou H,Zhang Q
Fri Feb 17 00:00:00 EST 2023
A Disintegrin and Metalloproteinase 10 (ADAM10) Is Essential for Oligodendrocyte Precursor Development and Myelination in the Mouse Brain.
Molecular neurobiology
Guo D,Huang F,Xue R,Ma Y,Xiao L,Lou H,Pan S
Wed Mar 01 00:00:00 EST 2023
Dulaglutide Improves Gliosis and Suppresses Apoptosis/Autophagy Through the PI3K/Akt/mTOR Signaling Pathway in Vascular Dementia Rats.
Neurochemical research
Guan T,Xiao Y,Xie X,Meng N,Qi Q,Xu J,Jiang X,Zhang Z,Teng Z,Lv P
Mon May 01 00:00:00 EDT 2023
Conditioned medium from bone marrow mesenchymal stem cells relieves spinal cord injury through suppression of Gal-3/NLRP3 and M1 microglia/macrophage polarization.
Pathology, research and practice
Wang Y,Wang X,Zou Z,Hu Y,Li S,Wang Y
Wed Mar 01 00:00:00 EST 2023
Anti-CHAC1 exosomes for nose-to-brain delivery of miR-760-3p in cerebral ischemia/reperfusion injury mice inhibiting neuron ferroptosis.
Journal of nanobiotechnology
Wang Y,Niu H,Li L,Han J,Liu Z,Chu M,Sha X,Zhao J
Mon Mar 27 00:00:00 EDT 2023
Transcriptome Profiles of IncRNA and mRNA Highlight the Role of Ferroptosis in Chronic Neuropathic Pain With Memory Impairment.
Frontiers in cell and developmental biology
Tang Y,Liu C,Zhu T,Chen H,Sun Y,Zhang X,Zhao Q,Wu J,Fei X,Ye S,Chen C
Sat Jul 16 00:00:00 EDT 2022
Neuroprotective Effects of Estrogen Through BDNF-Transient Receptor Potential Channels 6 Signaling Pathway in the Hippocampus in a Rat Model of Perimenopausal Depression.
Frontiers in aging neuroscience
Song Q,Huang W,Ye W,Yan H,Wang L,Yang Y,Cheng X,Zhang W,Zheng J,He P,He Y,Fang D,Han X
Tue Jul 26 00:00:00 EDT 2022
Fucoidan ameliorates LPS-induced neuronal cell damage and cognitive impairment in mice.
International journal of biological macromolecules
Wang Y,Wang Q,Duan L,Li X,Yang W,Huang T,Kong M,Guan F,Ma S
Thu Dec 01 00:00:00 EST 2022
Protective Effects of Bile Acids Against Hepatic Lipid Accumulation in Hybrid Grouper Fed a High-Lipid Diet.
Frontiers in nutrition
Xu J,Li X,Yao X,Xie S,Chi S,Zhang S,Cao J,Tan B
Sat Feb 12 00:00:00 EST 2022
Astrocyte-derived exosomal nicotinamide phosphoribosyltransferase (Nampt) ameliorates ischemic stroke injury by targeting AMPK/mTOR signaling to induce autophagy.
Cell death & disease
Deng Y,Duan R,Ding W,Gu Q,Liu M,Zhou J,Sun J,Zhu J
Tue Dec 20 00:00:00 EST 2022
Therapeutic effects of JLX001 on neuronal necroptosis after cerebral ischemia-reperfusion in rats.
Experimental brain research
Li W,Gou X,Xu D,Zhou L,Li F,Ye A,Hu Y,Li Y
Thu Dec 01 00:00:00 EST 2022
Downregulation of m6A Methyltransferase in the Hippocampus of Tyrobp -/- Mice and Implications for Learning and Memory Deficits.
Frontiers in neuroscience
Lv Z,Xu T,Li R,Zheng D,Li Y,Li W,Yang Y,Hao Y
Fri Apr 08 00:00:00 EDT 2022
TREM2 activation alleviates neural damage via Akt/CREB/BDNF signalling after traumatic brain injury in mice.
Journal of neuroinflammation
Yan J,Zhang Y,Wang L,Li Z,Tang S,Wang Y,Gu N,Sun X,Li L
Sat Dec 03 00:00:00 EST 2022
MFGE8 decreased neuronal apoptosis and neuroinflammation to ameliorate early brain injury induced by subarachnoid hemorrhage through the inhibition of HMGB1.
Human & experimental toxicology
Qiu J,Li W,Mu R,Wang L,Guo L,Ma L
Thu May 05 00:00:00 EDT 2022
SARM1 deletion in parvalbumin neurons is associated with autism-like behaviors in mice.
Cell death & disease
Xiang L,Wu Q,Sun H,Miao X,Lv Z,Liu H,Chen L,Gu Y,Chen J,Zhou S,Jiang H,Du S,Zhou Y,Dong H,Fan Y,Miao S,Lu Q,Chang L,Wang H,Lu Y,Xu X,Wang W,Huang Z
Fri Jul 22 00:00:00 EDT 2022
The cAMP Response Element- Binding Protein/Brain-Derived Neurotrophic Factor Pathway in Anterior Cingulate Cortex Regulates Neuropathic Pain and Anxiodepression Like Behaviors in Rats.
Frontiers in molecular neuroscience
Wen J,Xu Y,Yu Z,Zhou Y,Wang W,Yang J,Wang Y,Bai Q,Li Z
Wed Apr 13 00:00:00 EDT 2022
P2X7 Receptor-Induced Bone Cancer Pain by Regulating Microglial Activity via NLRP3/IL-1beta Signaling.
Pain physician
Wu P,Wu X,Zhou G,Wang Y,Liu X,Lv R,Liu Y,Wen Q
Tue Nov 01 00:00:00 EDT 2022
Sertad1 Induces Neurological Injury after Ischemic Stroke via the CDK4/p-Rb Pathway.
Molecules and cells
Li J,Li B,Bu Y,Zhang H,Guo J,Hu J,Zhang Y
Sat Apr 30 00:00:00 EDT 2022
The microbiota-gut-brain axis participates in chronic cerebral hypoperfusion by disrupting the metabolism of short-chain fatty acids.
Microbiome
Xiao W,Su J,Gao X,Yang H,Weng R,Ni W,Gu Y
Sun Apr 17 00:00:00 EDT 2022
Inhibition of Dectin-1 Alleviates Neuroinflammatory Injury by Attenuating NLRP3 Inflammasome-Mediated Pyroptosis After Intracerebral Hemorrhage in Mice: Preliminary Study Results.
Journal of inflammation research
Ding Z,Zhong Z,Wang J,Zhang R,Shao J,Li Y,Wu G,Tu H,Yuan W,Sun H,Wang Q
Tue Nov 01 00:00:00 EDT 2022
Repair of Spinal Cord Injury by Inhibition of PLK4 Expression Through Local Delivery of siRNA-Loaded Nanoparticles.
Journal of molecular neuroscience : MN
Gu Y,Zhang R,Jiang B,Xu X,Guan JJ,Jiang XJ,Zhou Y,Zhou YL,Chen X
Tue Mar 01 00:00:00 EST 2022
Apelin alleviated neuroinflammation and promoted endogenous neural stem cell proliferation and differentiation after spinal cord injury in rats.
Journal of neuroinflammation
Liu Q,Zhou S,Wang X,Gu C,Guo Q,Li X,Zhang C,Zhang N,Zhang L,Huang F
Mon Jun 20 00:00:00 EDT 2022
Lysosomal dysfunction is associated with NLRP3 inflammasome activation in chronic unpredictable mild stress-induced depressive mice.
Behavioural brain research
Li MM,Wang X,Chen XD,Yang HL,Xu HS,Zhou P,Gao R,Zhang N,Wang J,Jiang L,Liu N
Fri Aug 26 00:00:00 EDT 2022
Spinal cord astrocytes regulate myocardial ischemia-reperfusion injury.
Basic research in cardiology
Wu C,Liu R,Luo Z,Sun M,Qile M,Xu S,Jin S,Zhang L,Gross ER,Zhang Y,He S
Fri Nov 11 00:00:00 EST 2022
Ferroptosis participates in neuron damage in experimental cerebral malaria and is partially induced by activated CD8+ T cells.
Molecular brain
Liang J,Shen Y,Wang Y,Huang Y,Wang J,Zhu Q,Tong G,Yu K,Cao W,Wang Q,Li Y,Zhao Y
Mon Jun 20 00:00:00 EDT 2022
Upregulated hexokinase 2 expression induces the apoptosis of dopaminergic neurons by promoting lactate production in Parkinson's disease.
Neurobiology of disease
Li J,Chen L,Qin Q,Wang D,Zhao J,Gao H,Yuan X,Zhang J,Zou Y,Mao Z,Xiong Y,Min Z,Yan M,Wang CY,Xue Z
Tue Feb 01 00:00:00 EST 2022
Polystyrene micro- and nano-particle coexposure injures fetal thalamus by inducing ROS-mediated cell apoptosis.
Environment international
Yang D,Zhu J,Zhou X,Pan D,Nan S,Yin R,Lei Q,Ma N,Zhu H,Chen J,Han L,Ding M,Ding Y
Mon Aug 01 00:00:00 EDT 2022
Ras-related protein Rab-20 inhibition alleviates cerebral ischemia/reperfusion injury by inhibiting mitochondrial fission and dysfunction.
Frontiers in molecular neuroscience
Guo J,Zhang L,Bu Y,Li W,Hu J,Li J
Fri Nov 18 00:00:00 EST 2022
Pathogenic LRRK2 control of primary cilia and Hedgehog signaling in neurons and astrocytes of mouse brain.
eLife
Khan SS,Sobu Y,Dhekne HS,Tonelli F,Berndsen K,Alessi DR,Pfeffer SR
Mon Oct 18 00:00:00 EDT 2021
Enriched Environment Attenuates Pyroptosis to Improve Functional Recovery After Cerebral Ischemia/Reperfusion Injury.
Frontiers in aging neuroscience
Liu J,Zheng J,Xu Y,Cao W,Wang J,Wang B,Zhao L,Zhang X,Liao W
Fri Oct 15 00:00:00 EDT 2021
Genetic loss-of-function of activating transcription factor 3 but not C-type lectin member 5A prevents diabetic peripheral neuropathy.
Laboratory investigation; a journal of technical methods and pathology
Kan HW,Chang CH,Chang YS,Ko YT,Hsieh YL
Fri Oct 01 00:00:00 EDT 2021
Establishment and Evaluation of a Novel High-Efficiency Model of Graded Traumatic Brain Injury in Mice.
World neurosurgery
Chen B,Shi QX,Nie C,Zhao ZP,Luo L,Zhao QJ,Si SY,Xu BX,Wang T,Gao LY,Gu JW
Fri Oct 01 00:00:00 EDT 2021
Trans-10-hydroxy-2-decenoic acid protects against LPS-induced neuroinflammation through FOXO1-mediated activation of autophagy.
European journal of nutrition
You M,Miao Z,Tian J,Hu F
Thu Oct 01 00:00:00 EDT 2020