FIGURE: 1 / 1
Reconstitute at 0.2 mg/mL in sterile PBS.
ISL2 is a transcriptional factor that defines subclasses of motoneurons that segregate into columns in the spinal cord and select distinct axon pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Insulin gene enhancer protein ISL-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); Islet-2
Gene Aliases: ISL2
UniProt ID: (Human) Q96A47
Entrez Gene ID: (Human) 64843
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support