Immunogen sequence: MRNSYRFLA SSLSVVVSLL LIPEDVCEKI IGGNEVTPHS RPYMVLLSLD RKTICAGALI AKDWVLTAAH CNLNKRSQVI LGAHSITREE PTKQIMLVKK EFPYPCYDPA TREGDLKLLQ LTEKAKINKY VTILHLPKKG DDVKPGTMCQ VAGWGRTHNS ASWSDTLREV NITIIDRKVC NDRNHYNFNP VIGMNMVCAG SLRGGRDSCN GDSGSPLLCE GVFRGVTSFG LENKCGDPRG PGVYILLSKK HLNWIIMTIK GAV (1-262 aa encoded by BC015739)
Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface 'nonself' antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein described here is a T cell- and natural killer cell-specific serine protease that may function as a common component necessary for lysis of target cells by cytotoxic T lymphocytes and natural killer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autocrine thymic lymphoma granzyme-like serine protease; BLT esterase; CTL tryptase; CTLA-3; Cytotoxic T-lymphocyte proteinase 1; Cytotoxic T-lymphocyte-associated serine esterase-3; Fragmentin-1; granzyme 1; Granzyme A; Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factor serine protease); granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3); Granzyme-1; H factor; Hanukah factor; Hanukah factor serine protease); Hanukkah factor; HF; serine esterase 1; T cell-specific serine protease 1; TSP-1
Gene Aliases: AW494114; Ctla-3; CTLA3; GZMA; Hf; HFSP; Mtsp-1; SE1; TSP-1; TSP1
UniProt ID: (Human) P12544, (Mouse) P11032
Entrez Gene ID: (Human) 3001, (Mouse) 14938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support