Aliquoting is unnecessary for -20°C storage.
GSK3B is a proline-directed serine/threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation. It is a negative regulator of glucose homeostasis and is involved in energy metabolism, inflammation, ER-stress, mitochondrial dysfunction, and apoptotic pathways. GSK3B is a critical component of the Wnt/beta-catenin signaling pathway, and its activity results in the inhibition of Wnt signaling by mediating the degradation of beta-catenin. GSK3B is also involved in the Hedgehog, Notch, and insulin signaling pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.11.26; glycogen synthase kinase 3 beta variant 1; glycogen synthase kinase 3 beta variant 2; glycogen synthase kinase 3 beta variant 3; glycogen synthase kinase 3 beta variant 4; glycogen synthase kinase 3 beta variant 5; Glycogen synthase kinase-3 beta; GSK 3 beta; GSK 3B; GSK-3 beta; GSK-3 beta GSK3B; GSK-3b; GSK3 beta; GSK3beta isoform; Serine/threonine-protein kinase GSK3B
Gene Aliases: GSK3B; GSK3beta
UniProt ID: (Human) P49841
Entrez Gene ID: (Human) 2932, (Pig) 100126852
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support