Invitrogen
The PA1-73181 antibody reacts with Feline Leukemia virus p27.
PA1-73181 has been successfully used in ELISA applications.
The PA1-73181 antibody was raised against purified FeLv p27.
NOTE: Sodium azide is a potent inhibitor of peroxidase!
Feline leukemia virus (FeLV) is a retrovirus that infects cats. As a retrovirus, FeLV is transmitted as an RNA virus but the RNA is reverse transcribed into DNA. Retroviruses contradict the Central dogma of molecular biology. FeLV is usually transmitted between infected cats when the transfer of saliva or nasal secretions is involved. If not defeated by the animal’s immune system, the virus can be lethal. The disease caused by this virus is a form of cancer of blood cells called lymphocytes (a leukemia).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: FLV; FLV p27