Immunogen sequence: QVTAGRHGDD LRNTKQEIAE INRMIQRLRS EIDHVKKQCA NLQAAIADAE QRGEMALKDA KNKLEGLEDA LQKAKQDLAR LLKEYQELMN VKLALDVEIA TYRKLLEGEE CRLNGEGVGQ VNISVVQSTV SSGYGGASGV GSGLGLGGGS SYSYGSGLGV GGGFSSSSGR AIGGGLSSVG GGSSTIKYTT TSSSSRKSYK H
Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60-kDa keratin; CK-6A; CK-6B; CK-6C; CK-6D; CK-6E; cytokeratin 6A; cytokeratin 6B; cytokeratin 6C; cytokeratin 6D; Cytokeratin-6A; Cytokeratin-6B; Cytokeratin-6C; Cytokeratin-6D; Cytokeratin-6E; K6A; K6a keratin; K6B; K6C; kamp-keratin derived antimicrobial peptide; KDAMP; keratin 6A, , type II; keratin 6A, type II; keratin 6B, type II; keratin 6C, type II; keratin 6E; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6b; keratin complex 2, basic, gene 6c; keratin complex 2, gene 6a; keratin K6; Keratin K6h; keratin, epidermal type II, K6A; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 6B; Keratin, type II cytoskeletal 6C; Keratin-6-alpha; keratin-6-beta; Keratin-6A; Keratin-6B; Keratin-6C; keratin-like 1 (a type II keratin sequence); mK6-alpha; mK6-beta; Type-II keratin Kb10; Type-II keratin Kb12; Type-II keratin Kb6
Gene Aliases: 2310016L08Rik; BB144589; CK-6A; CK-6B; CK-6C; CK-6E; CK6A; CK6B; CK6C; CK6D; K6A; K6B; K6C; K6D; K6E; Ker2; Krt2-6; Krt2-6a; Krt2-6b; Krt2-6c; Krt6; KRT6A; KRT6B; KRT6C; KRT6D; KRT6E; KRTL1; mK6[a]; mK6[b]; MK6a; PC2; PC3; PC4; PPKNEFD
UniProt ID: (Human) P02538, (Human) P04259, (Human) P48668, (Mouse) P50446, (Rat) Q4FZU2
Entrez Gene ID: (Human) 3853, (Human) 3854, (Human) 286887, (Mouse) 16687, (Mouse) 16688, (Rat) 683313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support