Invitrogen
FIGURE: 1 / 1
This antibody reacts with native Cytochrome C, but does not recognize denatured cytochrome C. The antibody recognizes a region in the vicinity of amino acid 62 of rat Cytochrome C.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG17903-PA; CG17903-PB; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome C, expressed in somatic tissues; Cytochrome c, somatic; Cytochrome c-2; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; hc1; lethal (2) k13905; RH17228p
Gene Aliases: CG17903; CYC; CYC2; CYCS; CYCSA; Cyct-c-p; Cyt C; cyt c-p; Cyt-c; Cyt-c-p; Cyt-c2; cyt.c; CytC; CytC-2; Cytc-p; CYTC2; cytochrome c; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; HCS; l(2)k13905; pDMc01; THC4
UniProt ID: (Human) P99999, (Mouse) P62897, (Rat) P62898, (Fruit fly) P84029
Entrez Gene ID: (Human) 54205, (Mouse) 13063, (Rat) 25309, (Fruit fly) 34996, (Rabbit) 100341175, (Rabbit) 100357840
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support