Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
Description: This clone recognizes mouse CD294, also known as Prostaglandin D2 receptor 2 (PD2R2), G-protein coupled receptor 44 (GPR44) or Chemoattractant Receptor-Homologous Molecule Expressed On TH2 Cells (CRTH2). Although the clone works on unfixed mouse CD294-transfected cells, we recommend fixing and permeabilizing primary cells.
Applications Reported: This No3m1scz antibody has been reported for use in flow cytometric analysis.
Applications Tested: This No3m1scz antibody has been tested by flow cytometric analysis of mouse lysed whole blood cells using Protocol A: Two-step protocol for intracellular (cytoplasmic) proteins. This allows for the greatest flexibility for detection of surface and intracellular (cytoplasmic) proteins. Use of Protocol B: One-step protocol for intracellular (nuclear) proteins is recommended for staining of transcription factors in conjunction with surface and phosphorylated intracellular (cytoplasmic) proteins. Protocol C: Two-step protocol: Fixation/Methanol allows for the greatest discrimination of phospho-specific signaling between unstimulated and stimulated samples, but with limitations on the ability to stain specific surface proteins (refer to “Clone Performance Following Fixation/Permeabilization” located in the Best Protocols Section under the Resources tab online). All Protocols can be found in the “Staining Intracellular Antigens for Flow Cytometry Protocol” located in the Best Protocols Section under the Resources tab online. This may be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser
CRTH2 (GPR44) is a chemoattractant receptor-homologous molecule expressed on Th2 cells that is an identified receptor for the prostaglandin D2 (PGD2) and prostaglandin D receptor. CRTH2 is expressed on Th2 cells and eosinophils and mediates chemotaxis of these cells to PGD2 and is thus thought to be a key receptor mediating eosinophil and Th2 recruitment during allergic responses. However, CRTH2-null mice showed enhanced eosinophil recruitment into the lung consistent with observations that the CRTH2-null mice produced significantly higher amounts of interleukin-5 (IL-5) and IL-3. This suggests that CRTH2 plays a nonredundant role in restricting eosinophilia and allergic response in vivo. CRTH2 is also a novel member of the G protein-coupled leukocyte chemoattractant receptor family, which is selectively expressed in Th2 but not Th1 lineage cells. CRTH2 has been reported to be expressed in the central nervous system, peripheral tissues, and in blood cells, particularly activated T helper-2 cells. Two transcripts have been isolated: a 3.5 kb transcript expressed primarily in the thalamus, frontal cortex, pons and hippocampus; and a 3.4 kb transcript expressed in fetal liver, leukocytes and thymus. CRTH2 is selectively expressed in vivo in an activated state of Th2 cells including allergen-responsive Th2 cells, suggesting its pivotal roles in ongoing Th2-type immune reactions. At least two different isoforms of CRTH2 are known to exist. Single nucleotide polymorphisms in the 3' UTR of the CRTH2 gene have been associated with asthma susceptibility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD294; Chemoattractant receptor-homologous molecule expressed on Th2 cells; G protein-coupled receptor 44; G-protein coupled receptor 44; MGC130436; PGD2 receptor; Prostaglandin D2 receptor 2; prostaglandin D2 receptor CRTH2
Gene Aliases: Crth2; Gpr44; Grp45; Ptgdr2
UniProt ID: (Mouse) Q9Z2J6
Entrez Gene ID: (Mouse) 14764
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support