Product References
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Amyloid-beta and tau pathologies act synergistically to induce novel disease stage-specific microglia subtypes.
Molecular neurodegeneration
Kim DW,Tu KJ,Wei A,Lau AJ,Gonzalez-Gil A,Cao T,Braunstein K,Ling JP,Troncoso JC,Wong PC,Blackshaw S,Schnaar RL,Li T
14-1702-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to suggest that both Aβ and tau pathologies are required for the disease stage-specific induction of early-stage AD-Associated Microglia (EADAM) and late-stage AD-Associated Microglia (LADAM), and reveal Siglecs as biomarkers of Alzheimer's disease progression.
Sat Dec 17 00:00:00 EST 2022
Amyloid-beta and tau pathologies act synergistically to induce novel disease stage-specific microglia subtypes.
Molecular neurodegeneration
Kim DW,Tu KJ,Wei A,Lau AJ,Gonzalez-Gil A,Cao T,Braunstein K,Ling JP,Troncoso JC,Wong PC,Blackshaw S,Schnaar RL,Li T
14-1702-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to suggest that both Aβ and tau pathologies are required for the disease stage-specific induction of early-stage AD-Associated Microglia (EADAM) and late-stage AD-Associated Microglia (LADAM), and reveal Siglecs as biomarkers of Alzheimer's disease progression.
Sat Dec 17 00:00:00 EST 2022
Single-cell transcriptomics reveals distinct cell response between acute and chronic pulmonary infection of Pseudomonas aeruginosa.
MedComm
Hu X,Wu M,Ma T,Zhang Y,Zou C,Wang R,Zhang Y,Ren Y,Li Q,Liu H,Li H,Wang T,Sun X,Yang Y,Tang M,Li X,Li J,Gao X,Li T,Zhou X
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Immunohistochemistry
Thu Dec 01 00:00:00 EST 2022
Single-cell transcriptomics reveals distinct cell response between acute and chronic pulmonary infection of Pseudomonas aeruginosa.
MedComm
Hu X,Wu M,Ma T,Zhang Y,Zou C,Wang R,Zhang Y,Ren Y,Li Q,Liu H,Li H,Wang T,Sun X,Yang Y,Tang M,Li X,Li J,Gao X,Li T,Zhou X
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Immunohistochemistry
Thu Dec 01 00:00:00 EST 2022
Flow Cytometry and Mass Cytometry for Measuring the Immune Cell Infiltrate in Atherosclerotic Arteries.
Methods in molecular biology (Clifton, N.J.)
Orecchioni M,Meyer MA,Hedrick CC,Ley K
14-1702-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to describe the methodology used to isolate single immune cells from mouse aortas, followed by protocols for flow cytometry and mass cytometry for aortic immune cell characterization.
Fri Mar 04 00:00:00 EST 2022
Cells of the human intestinal tract mapped across space and time.
Nature
Elmentaite R,Kumasaka N,Roberts K,Fleming A,Dann E,King HW,Kleshchevnikov V,Dabrowska M,Pritchard S,Bolt L,Vieira SF,Mamanova L,Huang N,Perrone F,Goh Kai'En I,Lisgo SN,Katan M,Leonard S,Oliver TRW,Hook CE,Nayak K,Campos LS,Domínguez Conde C,Stephenson E,Engelbert J,Botting RA,Polanski K,van Dongen S,Patel M,Morgan MD,Marioni JC,Bayraktar OA,Meyer KB,He X,Barker RA,Uhlig HH,Mahbubani KT,Saeb-Parsy K,Zilbauer M,Clatworthy MR,Haniffa M,James KR,Teichmann SA
14-1702-82 was used in Flow Cytometry to comprehensively map the cellular landscape of the human intestinal tract from healthy paediatric and adult human gut.
Wed Sep 01 00:00:00 EDT 2021
Cells of the human intestinal tract mapped across space and time.
Nature
Elmentaite R,Kumasaka N,Roberts K,Fleming A,Dann E,King HW,Kleshchevnikov V,Dabrowska M,Pritchard S,Bolt L,Vieira SF,Mamanova L,Huang N,Perrone F,Goh Kai'En I,Lisgo SN,Katan M,Leonard S,Oliver TRW,Hook CE,Nayak K,Campos LS,Domínguez Conde C,Stephenson E,Engelbert J,Botting RA,Polanski K,van Dongen S,Patel M,Morgan MD,Marioni JC,Bayraktar OA,Meyer KB,He X,Barker RA,Uhlig HH,Mahbubani KT,Saeb-Parsy K,Zilbauer M,Clatworthy MR,Haniffa M,James KR,Teichmann SA
14-1702-82 was used in Flow Cytometry to comprehensively map the cellular landscape of the human intestinal tract from healthy paediatric and adult human gut.
Wed Sep 01 00:00:00 EDT 2021
Arf6 exacerbates allergic asthma through cell-to-cell transmission of ASC inflammasomes.
JCI insight
Lee S,Ishitsuka A,Kuroki T,Lin YH,Shibuya A,Hongu T,Funakoshi Y,Kanaho Y,Nagata K,Kawaguchi A
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Flow Cytometry
Mon Aug 23 00:00:00 EDT 2021
Evaluation of K18-hACE2 Mice as a Model of SARS-CoV-2 Infection.
The American journal of tropical medicine and hygiene
Moreau GB,Burgess SL,Sturek JM,Donlan AN,Petri WA,Mann BJ
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Eosinophils improve cardiac function after myocardial infarction.
Nature communications
Liu J,Yang C,Liu T,Deng Z,Fang W,Zhang X,Li J,Huang Q,Liu C,Wang Y,Yang D,Sukhova GK,Lindholt JS,Diederichsen A,Rasmussen LM,Li D,Newton G,Luscinskas FW,Liu L,Libby P,Wang J,Guo J,Shi GP
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Fibroblast-Derived STC-1 Modulates Tumor-Associated Macrophages and Lung Adenocarcinoma Development.
Cell reports
Kamata T,So TY,Ahmed Q,Giblett S,Patel B,Luo J,Reddel R,Pritchard C
14-1702 was used in Flow cytometry/Cell sorting to identify the pleiotropic glycoprotein stanniocalcin-1 (STC1) as a regulator of TAM-TAF interactions.
Tue Jun 23 00:00:00 EDT 2020
Pro-inflammatory Aorta-Associated Macrophages Are Involved in Embryonic Development of Hematopoietic Stem Cells.
Immunity
Mariani SA,Li Z,Rice S,Krieg C,Fragkogianni S,Robinson M,Vink CS,Pollard JW,Dzierzak E
14-1702-82 was used in Immunohistochemistry (Frozen) to show here that macrophage appearance in the AGM was dependent on the chemokine receptor Cx3cr1.
Tue Jun 18 00:00:00 EDT 2019
ILC2 transfers to apolipoprotein E deficient mice reduce the lipid content of atherosclerotic lesions.
BMC immunology
Mantani PT,Dunér P,Ljungcrantz I,Nilsson J,Björkbacka H,Fredrikson GN
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Immunohistochemistry
Tue Dec 10 00:00:00 EST 2019
Treating ischemia via recruitment of antigen-specific T cells.
Science advances
Kwee BJ,Seo BR,Najibi AJ,Li AW,Shih TY,White D,Mooney DJ
14-1702-82 was used in Immunohistochemistry to support the premise that antigen depots may provide a treatment for ischemic diseases in patients previously vaccinated with aluminum-containing adjuvants.
Mon Jul 01 00:00:00 EDT 2019
IL-27 targets Foxp3+ Tregs to mediate antiinflammatory functions during experimental allergic airway inflammation.
JCI insight
Nguyen QT,Jang E,Le HT,Kim S,Kim D,Dvorina N,Aronica MA,Baldwin WM,Asosingh K,Comhair S,Min B
14-1702 was used in Flow cytometry/Cell sorting to discover that Tregs during allergic airway inflammation are the primary target cells of IL-27 in order to mediate antinflammatory functions.
Thu Jan 24 00:00:00 EST 2019
11 Beta-hydroxysteroid dehydrogenase type 1 regulates synovitis, joint destruction, and systemic bone loss in chronic polyarthritis.
Journal of autoimmunity
Hardy RS,Fenton C,Croft AP,Naylor AJ,Begum R,Desanti G,Buckley CD,Lavery G,Cooper MS,Raza K
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Functional characterization of oxazolone-induced colitis and survival improvement by vagus nerve stimulation.
PloS one
Meroni E,Stakenborg N,Gomez-Pinilla PJ,De Hertogh G,Goverse G,Matteoli G,Verheijden S,Boeckxstaens GE
14-1702-82 was used in Flow Cytometry to characterise the immune response in oxazolone-induced colitis and study the therapeutic effect of vagus nerve stimulation.
Tue Nov 27 00:00:00 EST 2018
Der f 31, a novel allergen from Dermatophagoides farinae, activates epithelial cells and enhances lung-resident group 2 innate lymphoid cells.
Scientific reports
Wang H,Lin J,Zeng L,Ouyang C,Ran P,Yang P,Liu Z
Published figure using CD170 (Siglec F) monoclonal antibody (Product # 14-1702-82) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017