FIGURE: 1 / 1
This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activation induced deaminase; Activation-induced cytidine deaminase; AID; Cytidine aminohydrolase; epididymis secretory protein Li 284; integrated into Burkitt's lymphoma cell line Ramos; Single-stranded DNA cytosine deaminase
Gene Aliases: AICDA; AID; ARP2; CDA2; HEL-S-284; HIGM2
UniProt ID: (Human) Q9GZX7, (Mouse) Q9WVE0
Entrez Gene ID: (Human) 57379, (Mouse) 11628
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support