Proteintech
Immunogen sequence: MERASLIQK AKLAEQAERY EDMAAFMKGA VEKGEELSCE ERNLLSVAYK NVVGGQRAAW RVLSSIEQKS NEEGSEEKGP EVREYREKVE TELQGVCDTV LGLLDSHLIK EAGDAESRVF YLKMKGDYYR YLAEVATGDD KKRIIDSARS AYQEAMDISK KEMPPTNPIR LGLALNFSVF HYEIANSPEE AISLAKTTFD EAMADLHTLS EDSYKDSTLI MQLLRDNLTL WTADNAGEEG GEAPQEPQS (1-248 aa encoded by BC002995 )
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14-3-3 protein sigma; 14-3-3 sigma; 14-3-3sigma; Epithelial cell marker protein 1; HGNC:10773; Stratifin
Gene Aliases: HME1; SFN; YWHAS
UniProt ID: (Human) P31947
Entrez Gene ID: (Human) 2810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support